Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ USP9X Monoclonal Antibody (OTI2G7)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526367
Description
USP9X Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
USP9X is an ubiquitin-specific protease involved in the removal and recycling of ubiquitin from proteins degraded in proteasomes. As a deubiquitinating enzyme, USP9x has a critical function in tight junction biogenesis by controlling the levels of the exchange factor for Arf6 (EFA6), a protein shown to facilitate tight junction formation during a narrow temporal window preceding the establishment of cell polarity. USP9X is also located on the X-chromosome but escapes X-inactivation, and defects in USP9X may cause gonadal degeneration in Turner syndrome.
Specifications
USP9X | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P70398, Q93008 | |
USP9X | |
Human recombinant protein fragment corresponding to amino acids 2246-2570 of USP9X produced in E.coli. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
OTI2G7 | |
Unconjugated | |
USP9X | |
5730589N07Rik; AA407302; AA407699; AL022658; AL022749; Deubiquitinating enzyme FAF-X; DFFRX; Drosophila fat facets related, X-linked; FAF; Fafl; FAF-X; FAM; fat facets homolog; Fat facets in mammals; fat facets protein related, X-linked; fat facets protein-related, X-linked; fats facets protein related, X; hFAM; MRX99; MRXS99F; Probable ubiquitin carboxyl-terminal hydrolase FAF-X; putative ubiquitin carboxyl-terminal hydrolase FAF-X; putative ubiquitin carboxyl-terminal hydrolase FAF-x-like protein; RGD1560056; Ubiquitin carboxyl-terminal hydrolase FAM; ubiquitin specific peptidase 9 X-linked; ubiquitin specific peptidase 9, X chromosome; ubiquitin specific peptidase 9, X-linked; ubiquitin specific protease 9 SSSRF- isoform; ubiquitin specific protease 9, X chromosome; ubiquitin specific protease 9, X chromosome (fat facets-like Drosophila); ubiquitin specific protease 9, X-linked; ubiquitin thioesterase FAF-X; ubiquitin thiolesterase FAF-X; ubiquitin-specific processing protease FAF-X; ubiquitin-specific protease 9, X chromosome; Ubiquitin-specific-processing protease FAF-X; USP9; USP9X | |
Mouse | |
Affinity chromatography | |
RUO | |
22284, 363445, 480885, 8239 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction