Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USP9Y Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570428
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570428 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570428 Supplier Invitrogen™ Supplier No. PA570428
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Dog: 86%; Horse: 86%; Human: 100%; Mouse: 86%; Pig: 86%; Rabbit: 86%; Rat: 86%.

USP9Y gene ontology annotations related to this gene include protein deubiquitination; ubiquitin-dependent protein catabolic process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP9Y
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene USP9Y
Gene Accession No. O00507
Gene Alias Deubiquitinating enzyme FAF-Y; DFFRY; Fafl2; Fat facets protein-related, Y-linked; probable ubiquitin carboxyl-terminal hydrolase FAF-X; probable ubiquitin carboxyl-terminal hydrolase FAF-Y; SPGFY2; ubiquitin specific peptidase 9, Y chromosome; ubiquitin specific peptidase 9, Y-linked; ubiquitin specific peptidase 9, Y-linked (fat facets-like, Drosophila); ubiquitin specific protease 9, Y chromosome; ubiquitin thioesterase FAF-Y; ubiquitin thiolesterase FAF-Y; ubiquitin-specific processing protease FAF-Y; ubiquitin-specific protease 9, Y chromosome; Ubiquitin-specific-processing protease FAF-Y; USP9Y
Gene Symbols USP9Y
Host Species Rabbit
Immunogen synthetic peptide directed towards the C terminal region of human USP9Y.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8287
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.