Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UST Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542905
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542905 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542905 Supplier Invitrogen™ Supplier No. PA542905
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: YFKGVLSIYK DPEHRKLGNM TVTVKKTVPS PEAVQILYQR MRYEYEFYHY Sequence homology: Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

Uronyl 2-sulfotransferase transfers sulfate to the 2-position of uronyl residues, such as iduronyl residues in dermatan sulfate and glucuronyl residues in chondroitin sulfate (Kobayashi et al., 1999 [PubMed 10187838]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen UST
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene UST
Gene Accession No. Q9Y2C2
Gene Alias 2OST; D930010O20Rik; dermatan/chondroitin sulfate 2-sulfotransferase; DS2ST; UA2OST; Uronyl 2-sulfotransferase; uronyl-2-sulfotransferase; UST
Gene Symbols UST
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human UST (aa 301-350).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10090
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.