Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UST Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556902
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556902 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556902 Supplier Invitrogen™ Supplier No. PA556902
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YNRVGKCGSR TVVLLLRILS EKHGFNLVTS DIHNKTRLTK NEQMELIKNI STAEQPYLFT RHVHFLNFSR FG Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 96%.

Uronyl 2-sulfotransferase transfers sulfate to the 2-position of uronyl residues, such as iduronyl residues in dermatan sulfate and glucuronyl residues in chondroitin sulfate (Kobayashi et al., 1999 [PubMed 10187838]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen UST
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UST
Gene Accession No. Q9Y2C2
Gene Alias 2OST; D930010O20Rik; dermatan/chondroitin sulfate 2-sulfotransferase; DS2ST; UA2OST; Uronyl 2-sulfotransferase; uronyl-2-sulfotransferase; UST
Gene Symbols UST
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UST. Recombinant protein control fragment (Product #RP-95607).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10090
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.