Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ustilago sphaerogena RNU2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117624
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117624 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117624 Supplier Invitrogen™ Supplier No. PA5117624
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

RNase U2 is a 114aa endoribonuclease secreted by the fungus Ustilago sphaerogena. RNase U2 displays optimum pH for enzymatic activity at pH 3.5-5.5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ustilago sphaerogena RNU2
Applications Western Blot
Classification Polyclonal
Concentration 3.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Alias Ribonuclease U2; RNase U2
Host Species Rabbit
Immunogen Recombinant Ustilago sphaerogena Ribonuclease U2 protein (1-114aa).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Fungi
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.