Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UT-A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116409
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116409 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116409 Supplier Invitrogen™ Supplier No. PA5116409
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UT-A1 Polyclonal Antibody for Western Blot, IHC (P)

UT-A1, a kidney-specific urea transporter is expressed in the renal collecting duct where it mediates trans-epithelial urea transport and is a target for regulation by vasopressin. Urea movement out of the collecting duct in the inner medulla of the kidney allows accumulation of urea in the medullary interstitium, thereby allowing maximum water reabsorption from the collecting ducts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UT-A1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Slc14a2
Gene Accession No. Q15849
Gene Alias FLJ16167; HUT2; hUT-A6; MGC119566; MGC119567; plasma membrane urea transporter; SLC14A2; Slc14a2_v4; Slc14a2T; solute carrier family 14 (urea transporter), member 2; solute carrier family 14 (urea transporter), member 2, variant 4; solute carrier family 14 (urea transporter), member 2T; solute carrier family 14 member 2; solute carrier family 14, member 2; testName; testSymbol; urea transport protein; urea transporter 2; Urea transporter, kidney; urea transporter-2; UrT1-C; UrT1-D; UT1; UT2; UTA; UTA1; UT-A1; UT-A2; UT-A3; UTR
Gene Symbols Slc14a2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 42-76 amino acids from the human region of human SLC14A2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8170
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.