Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UT-A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577786
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577786 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577786 Supplier Invitrogen™ Supplier No. PA577786
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

1 μg/mL of PA5-77786 was sufficient for detection of UT-A1 in 20 μg of rat kidney tissue lysate by colorimetric immunoblot analysis using Goat anti-rabbit IgG:HRP as the secondary antibody. Detects approximately 97 and 127kDa.

UT-A1, a kidney-specific urea transporter is expressed in the renal collecting duct where it mediates trans-epithelial urea transport and is a target for regulation by vasopressin. Urea movement out of the collecting duct in the inner medulla of the kidney allows accumulation of urea in the medullary interstitium, thereby allowing maximum water reabsorption from the collecting ducts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UT-A1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Gene Slc14a2
Gene Accession No. Q62668, Q8R4T9
Gene Alias FLJ16167; HUT2; hUT-A6; MGC119566; MGC119567; plasma membrane urea transporter; SLC14A2; Slc14a2_v4; Slc14a2T; solute carrier family 14 (urea transporter), member 2; solute carrier family 14 (urea transporter), member 2, variant 4; solute carrier family 14 (urea transporter), member 2T; solute carrier family 14 member 2; solute carrier family 14, member 2; testName; testSymbol; urea transport protein; urea transporter 2; Urea transporter, kidney; urea transporter-2; UrT1-C; UrT1-D; UT1; UT2; UTA; UTA1; UT-A1; UT-A2; UT-A3; UTR
Gene Symbols Slc14a2
Host Species Rabbit
Immunogen Produced against a synthetic peptide mapped to the C-terminal tail (amino acids 911-929) of rat UT-A1 (antibody designation L194).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27411, 54302
Target Species Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.