Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UT2/SLC14A2 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP144375AF350
Description
UT2/SLC14A2 Polyclonal antibody specifically detects UT2/SLC14A2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
UT2/SLC14A2 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
FLJ16167, HUT2UT-A2, hUT-A6, MGC119566, solute carrier family 14 (urea transporter), member 2, Solute carrier family 14 member 2, urea transporter 2, Urea transporter, kidney, urea transporter-2, UT2MGC119567, UTA, UTR | |
Produced against a synthetic peptide mapped to the C-terminal tail (amino acids 911-929) of rat UT-A1 (antibody designation L194) | |
0.1 mL | |
ABC Transporters, Neuroscience | |
8170 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction