Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uteroglobin Goat anti-Mouse, Polyclonal, MilliporeSigma™

Catalog No. ABS1673MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS1673MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1673MI Supplier MilliporeSigma Supplier No. ABS1673
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Detect Uteroglobin using this goat polyclonal Anti-Uteroglobin Antibody immunohistochemistry, and Western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Uteroglobin
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Scgb1a1,Cc10,Ugb,Utg
Host Species Goat
Immunogen His-tagged full length recombinant rat uteroglobin.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_037183
Test Specificity This polyclonal antibody detects uteroglobin in mouse and rat tissues.
Target Species Mouse
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.