Learn More
Invitrogen™ Uteroglobin Monoclonal Antibody (3A8B8)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA517170
Description
MA5-17170 targets SCGB1A1 in FACS, indirect ELISA, and WB applications and shows reactivity with Human samples. The MA5-17170 immunogen is purified recombinant fragment of human SCGB1A1 (amino acids: 26-91) expressed in E. Coli. MA5-17170 detects SCGB1A1 which has a predicted molecular weight of approximately 10kDa.
Uteroglobin, which is a member of the Secretoglobin superfamily and is also known as Clara cell phospholipid-binding protein, is a multifunctional protein that can exert anti-inflammatory and anti-tumorigenic effects by binding small hydrophobic molecules such as phospholipids and prostaglandins. The small, non-glycosylated protein named for its high levels of expression in pre-implantation embryos, where it exhibits growth stimulatory effects, is produced and secreted by the non-ciliated, non-mucous Clara cells predominant in the epithelial surfaces of pulmonary airways, as well as other non-ciliated epithelia. Members of the Secretoglobin superfamily demonstrate a high level of structural conservation and are characterized as small, secretory homo- or heterodimers. In addition to sequestering pro-inflammatory mediators and carcinogens, Uteroglobin has been implicated in the inhibition of cell migration and invasion, platelet aggregation, and T cell differentiation.
Specifications
Uteroglobin | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P11684 | |
SCGB1A1 | |
Purified recombinant fragment of human SCGB1A1 (amino acids: 26-91) expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Western Blot | |
3A8B8 | |
Unconjugated | |
SCGB1A1 | |
Blastokinin; CC10; CC16; CCPBP; CCSP; Clara cell 17 kDa protein; clara cell phospholipid-binding protein; clara cell secretory protein; clara cells 10 kDa secretory protein; PCBB; PCB-binding protein; PCB-BP; SCGB1A1; Secretoglobin family 1A member 1; secretoglobin, family 1A, member 1 (uteroglobin); UG; Ugb; UP1; UP-1; Urinary protein 1; urine protein 1; uteroglobin; Uteroglobin (Clara cell secretory protein); Utg | |
Mouse | |
Protein G | |
RUO | |
7356 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.