Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Uteroglobin Polyclonal Antibody
Uteroglobin Polyclonal Antibody
Supplier: Thermo Scientific PA579957
Description
Uteroglobin Polyclonal Antibody for Western Blot, IHC (P)

Specifications
Uteroglobin | |
Polyclonal | |
Unconjugated | |
SCGB1A1 | |
Blastokinin; CC10; CC16; CCPBP; CCSP; Clara cell 17 kDa protein; clara cell phospholipid-binding protein; clara cell secretory protein; clara cells 10 kDa secretory protein; PCBB; PCB-binding protein; PCB-BP; SCGB1A1; Secretoglobin family 1A member 1; secretoglobin, family 1A, member 1 (uteroglobin); UG; Ugb; UP1; UP-1; Urinary protein 1; urine protein 1; uteroglobin; Uteroglobin (Clara cell secretory protein); Utg | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25575 | |
-20°C | |
Lyophilized |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 4mg trehalose and 0.05mg sodium azide | |
P17559 | |
SCGB1A1 | |
E. coli-derived rat Uteroglobin recombinant protein (Position: S20-V96). | |
100 μg | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction