Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeproTech Uteroglobin Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Catalog No. 500P330BT50
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
500P330BT50 50 μg
50-271-8500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 500P330BT50 Supplier PeproTech Supplier No. 500P330BT50UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: MEICPSFQRV IETLLMDTPS SYEAAMELFS PDQDMREAGA QLKKLVDTLP QKPRESIIKL MEKIAQSSLC N. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Uteroglobin. Anti-Human Uteroglobin-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human Uteroglobin by sandwich ELISA (using 100 μL/well) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human Uteroglobin (500-P330) as a capture antibody, allows the detection of at least 2000-4000 pg/mL of Recombinant Human Uteroglobin. Western Blot: To detect Human Uteroglobin by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Human Uteroglobin is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P330BT-1MG will be provided as 2 x 500 μg

Uteroglobin, which is a member of the Secretoglobin superfamily and is also known as Clara cell phospholipid-binding protein, is a multifunctional protein that can exert anti-inflammatory and anti-tumorigenic effects by binding small hydrophobic molecules such as phospholipids and prostaglandins. The small, non-glycosylated protein named for its high levels of expression in pre-implantation embryos, where it exhibits growth stimulatory effects, is produced and secreted by the non-ciliated, non-mucous Clara cells predominant in the epithelial surfaces of pulmonary airways, as well as other non-ciliated epithelia. Members of the Secretoglobin superfamily demonstrate a high level of structural conservation and are characterized as small, secretory homo- or heterodimers. In addition to sequestering pro-inflammatory mediators and carcinogens, Uteroglobin has been implicated in the inhibition of cell migration and invasion, platelet aggregation, and T cell differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Uteroglobin
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Biotin
Formulation PBS with no preservative
Gene SCGB1A1
Gene Accession No. P11684
Gene Alias Blastokinin; CC10; CC16; CCPBP; CCSP; Clara cell 17 kDa protein; clara cell phospholipid-binding protein; clara cell secretory protein; clara cells 10 kDa secretory protein; PCBB; PCB-binding protein; PCB-BP; SCGB1A1; Secretoglobin family 1A member 1; secretoglobin, family 1A, member 1 (uteroglobin); UG; Ugb; UP1; UP-1; Urinary protein 1; urine protein 1; uteroglobin; Uteroglobin (Clara cell secretory protein); Utg
Gene Symbols SCGB1A1
Host Species Rabbit
Immunogen E.coli-derived Recombinant Human Uteroglobin.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7356
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.