Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uteroglobin Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986128
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6128 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6128 Supplier Bioss Supplier No. BS4684R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Uteroglobin encodes a member of the secretoglobin family of small secreted proteins. The encoded protein has been implicated in numerous functions including anti-inflammation, inhibition of phospholipase A2 and the sequestering of hydrophobic ligands. Defects in this gene are associated with a susceptibility to asthma.

Specifications

Antigen Uteroglobin
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SCGB1A1
Gene Accession No. P11684
Gene Alias Blastokinin; CC10; CC16; CCPBP; CCSP; Clara cell 17 kDa protein; Clara cell phospholipid-binding protein; clara cell secretory protein; Clara cells 10 kDa secretory protein; PCBB; PCB-binding protein; PCB-BP; Scgb1a1; Secretoglobin family 1A member 1; secretoglobin, family 1A, member 1 (uteroglobin); UG; Ugb; UP1; UP-1; Urinary protein 1; urine protein 1; Uteroglobin; Uteroglobin (Clara cell secretory protein); Utg
Gene Symbols SCGB1A1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from mouse Uteroglobin.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7356
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.