Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Uteroglobin/SCGB1A1 Antibody (030), DyLight 550, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289649R
Description
Uteroglobin/SCGB1A1 Monoclonal antibody specifically detects Uteroglobin/SCGB1A1 in Human samples. It is validated for ELISASpecifications
Uteroglobin/SCGB1A1 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
030 | |
ELISA | |
blastokinin, CC10Urine protein 1, CC16, CCSPCCPBP, Clara cell phospholipid-binding protein, Clara cells 10 kDa secretory protein, Secretoglobin family 1A member 1, secretoglobin, family 1A, member 1 (uteroglobin), UGBUP-1, UP1, Urinary protein 1, uteroglobin | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Uteroglobin/SCGB1A1 (P11684; Met1-Asn91). | |
0.1 mL | |
Biologically Active Proteins, Cytokine Research | |
7356 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction