Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Uteroglobin/SCGB1A1 Antibody (122), Allophycocyanin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290517APC
Description
Uteroglobin/SCGB1A1 Monoclonal antibody specifically detects Uteroglobin/SCGB1A1 in Mouse samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Uteroglobin/SCGB1A1 | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Immunohistochemistry (Paraffin) | |
122 | |
Immunohistochemistry-Paraffin | |
blastokinin, CC10Urine protein 1, CC16, CCSPCCPBP, Clara cell phospholipid-binding protein, Clara cells 10 kDa secretory protein, Secretoglobin family 1A member 1, secretoglobin, family 1A, member 1 (uteroglobin), UGBUP-1, UP1, Urinary protein 1, uteroglobin | |
Rabbit immunized with purified, recombinant Mouse Uteroglobin/SCGB1A1 (Uniprot#: Q06318, Met1-Phe96) | |
0.1 mL | |
Biologically Active Proteins, Cytokine Research | |
7356 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction