Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UTF1 Monoclonal Antibody (5B6)
GREENER_CHOICE

Catalog No. PIMA517197
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517197 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517197 Supplier Invitrogen™ Supplier No. MA517197
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17197 targets UTF1 in FACS, indirect ELISA, and WB applications and shows reactivity with Human samples. The MA5-17197 immunogen is purified recombinant fragment of human UTF1 (amino acids: 148-214) expressed in E. Coli. MA5-17197 detects UTF1 which has a predicted molecular weight of approximately 36.4kDa.

Undifferentiated transcription factor-1 (UTF-1) is used as marker for the undifferentiated state of pluripotent stem cells.UTF1 is a chromatin-associated protein with core histone-like properties. UTF1 further acts as a transcriptional repressor and is required for proper differentiation of pluripotent cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UTF1
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 5B6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene UTF1
Gene Accession No. Q5T230
Gene Alias AI505934; OTTHUMP00000020786; Undifferentiated embryonic cell transcription factor 1; UTF1
Gene Symbols UTF1
Host Species Mouse
Immunogen Purified recombinant fragment of human UTF1 (amino acids: 148-214) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8433
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.