Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UTF1 Monoclonal Antibody (E0-F4)
GREENER_CHOICE

Catalog No. PIMA544624
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544624 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544624 Supplier Invitrogen™ Supplier No. MA544624
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

UTF1 Monoclonal Antibody for Western Blot, ICC/IF

Undifferentiated transcription factor-1 (UTF-1) is used as marker for the undifferentiated state of pluripotent stem cells.UTF1 is a chromatin-associated protein with core histone-like properties. UTF1 further acts as a transcriptional repressor and is required for proper differentiation of pluripotent cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UTF1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone E0-F4
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene UTF1
Gene Accession No. Q5T230, Q6J1H4
Gene Alias AI505934; OTTHUMP00000020786; Undifferentiated embryonic cell transcription factor 1; UTF1
Gene Symbols UTF1
Host Species Mouse
Immunogen Synthetic peptide within Human UTF1 aa 51-100/341.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22286, 681901, 8433
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.