Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UTF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540319
Description
Peptide sequence: AREDPDSPPG RPEDCAPPPA APPSLNTALL QTLGHLGDIA NILGPLRDQL Sequence homology: Cow: 93%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Mouse: 86%; Pig: 93%; Rat: 86%.
Undifferentiated transcription factor-1 (UTF-1) is used as marker for the undifferentiated state of pluripotent stem cells.UTF1 is a chromatin-associated protein with core histone-like properties. UTF1 further acts as a transcriptional repressor and is required for proper differentiation of pluripotent cells.
Specifications
| UTF1 | |
| Polyclonal | |
| Unconjugated | |
| UTF1 | |
| AI505934; OTTHUMP00000020786; Undifferentiated embryonic cell transcription factor 1; UTF1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 8433 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5T230 | |
| UTF1 | |
| Synthetic peptide directed towards the middle region of human UTF1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction