Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIOTIX™ uTIP™ CleanPak™ Reload, Low-Retention, Non-Filter Universal Pipette Tips, Standard
GSA_VAGREENER_CHOICE

Catalog No. 12111024
Click to view available options
Volume (Metric):
10 μL
1250 μL
200 μL
300 μL

Designed with hermetically sealed bag to avoid contamination. Biotix™ uTIP™ CleanPak™ Reload, Low-Retention, Non-Filter Universal Pipette Tips reduce time to rack products, cost compared to pre-racked products, and environmental footprint.

  • Fits all standard pipettes
  • Improves ergonomics by lowering insertion and ejection forces with FlexFit and Blade technologies
  • CleanPak reload features advanced technology of Biotix tips
  • Allows re-use of previous racks
  • 960 tips on lab bench using the same surface area as 1 rack
  • Sterile and non-Sterile re-sealable bag available
  • Clear bag and color coded snap cards for quick identification
  • Fully recyclable and contains no contaminating cardboard or paper
  • Inserts are easy to transfer to Biotix empty racks
  • All racks include label with unique bar-coded serial number, part number, and description for traceability to individual rack

Certifications

ISO 9001 , FDA Class I

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 300 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Compatibility Universal
Format Refill
Tip Style Fine Point
Product Type Pipette Tips
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line uTIP CleanPak
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.