Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIOTIX™ uTIP™ Low Retention Non-Filter Universal Pipette Tips, Standard, Bulk
GSA_VAGREENER_CHOICE

Catalog No. p-7075321
Click to view available options
Volume (Metric):
10 μL
1250 μL
200 μL
300 μL

Packaged in re-sealable bags making storage and racking simple. Biotix™ uTIP™ Low Retention Non-Filter Universal Pipette Tips, Bulk are RNase, DNase, Pyrogens, and PCR Inhibitors free.

  • Fits leading pipette brands and improves ergonomics by lowering insertion and ejection forces
  • Achieved through patented technologies including FlexFit and Blade
  • Contain X-Resin giving uniform low retention properties
  • Results in minimal sample retention in tip improving accuracy and precision of results
  • Re-sealable bag
  • Tips must be racked by hand
  • Sterile products undergo E-Beam radiation

Certifications

ISO 9001 , FDA Class I

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 10 μL
Filtered Non-filtered
Color Clear
Autoclavable Autoclavable
Compatibility Universal
Format Bulk
Tip Style Fine Point
Product Type Pipette Tips
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line uTip
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.