Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BIOTIX™ uTIP™ Sterile Low Retention Universal Pipette Tips, Standard

Shop All Biotix Products


Click to view available options
Volume (Metric):
250 μL
1000 μL
Description
Certifications
ISO 9001
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications
Specifications
Description | 250 μL, Racked, Sterile |
Volume (Metric) | 250 μL |
Filtered | Non-filtered |
Sterility | Sterile |
Color | Clear |
Rack Color | Green |
Autoclavable | Autoclavable |
Model | Universal |
Length (Metric) | 59.54 mm |
Certifications/Compliance | ISO 9001 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction