Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UTP11L Antibody (OTI5F3), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274823F
Description
UTP11L Monoclonal specifically detects UTP11L in Human samples. It is validated for Immunohistochemistry.Specifications
UTP11L | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
UTP11L | |
Full length recombinant protein of human UTP11L (NP_057121) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Immunohistochemistry | |
OTI5F3 | |
Immunohistochemistry | |
CGI94, CGI-94, probable U3 small nucleolar RNA-associated protein 11, U3 snoRNA-associated protein 11, UTP11-like protein, UTP11-like, U3 small nucleolar ribonucleoprotein, (yeast) | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication, Cytokine Research | |
51118.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction