Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UTP11L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561100
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561100 Supplier Invitrogen™ Supplier No. PA561100
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HIIKETKEEV TPEQLKLMRT QDVKYIEMKR VAEAKKIERL KSELHLLDFQ GKQQNKHVFF FDTKKEVEQF DVATHLQTAP EL Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.

Involved in nucleolar processing of pre-18S ribosomal RNA. Target protein may have a role in maintainance of neuronal function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UTP11L
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UTP11
Gene Accession No. Q9Y3A2
Gene Alias 2700082D03Rik; Cgi94; CGI-94; HDCMB12P; Probable U3 small nucleolar RNA-associated protein 11; RGD1559610; U3 snoRNA-associated protein 11; UTP11; UTP11, small subunit processome component homolog (S. cerevisiae); UTP11L; UTP11-like protein; UTP11-like, U3 small nucleolar ribonucleoprotein; UTP11-like, U3 small nucleolar ribonucleoprotein, (yeast)
Gene Symbols UTP11
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UTP11L. Recombinant protein control fragment (Product #RP-94494).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51118
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.