Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UTP11L Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | UTP11L |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
UTP11L Polyclonal specifically detects UTP11L in Human samples. It is validated for Western Blot.Specifications
UTP11L | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication, Cytokine Research | |
PBS buffer, 2% sucrose | |
51118 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
CGI94, CGI-94, probable U3 small nucleolar RNA-associated protein 11, U3 snoRNA-associated protein 11, UTP11-like protein, UTP11-like, U3 small nucleolar ribonucleoprotein, (yeast) | |
The immunogen is a synthetic peptide directed towards the N-terminal region of Human UTP11LL (NP_057121). Peptide sequence AKSRQREHRERSQPGFRKHLGLLEKKKDYKLRADDYRKKQEYLKALRKKA | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title