Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UTP14A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101250
Description
Antibody detects endogenous levels of total UTP14A.
UTP14A, a related protein, may also be required for ribosome biogenesis, but not necessarily in a male-specific manner. The gene encoding UTP14C maps to human chromosome 13, which houses over 400 genes, such as BRCA2 and RB1, and comprises nearly 4% of the human genome. As with most chromosomes, polysomy of part or all of chromosome 13 is deleterious to development and decreases the odds of survival. Trisomy 13, also known as Patau syndrome, is deadly and the few who survive past one year suffer from permanent neurologic defects, difficulty eating and vulnerability to serious respiratory infections.
Specifications
UTP14A | |
Polyclonal | |
Unconjugated | |
UTP14A | |
2700066J21Rik; Antigen NY-CO-16; dJ537K23.3; Jsd-like X-linked protein; JsdX; Juvenile spermatogonial depletion-like X-linked protein; Kiaa0266; mKIAA0266; NYCO16; NY-CO-16; SDCCAG16; Serologically defined colon cancer antigen 16; T25628; U3 small nucleolar RNA-associated protein 14 homolog A; UTP14, U3 small nucleolar ribonucleoprotein, homolog A; UTP14, U3 small nucleolar ribonucleoprotein, homolog A (yeast); UTP14A; UTP14A small subunit (SSU) processome component; UTP14A small subunit processome component | |
Rabbit | |
Affinity chromatography | |
RUO | |
10813, 72554 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q640M1, Q9BVJ6 | |
UTP14A | |
A synthesized peptide derived from human UTP14A(Accession Q9BVJ6), corresponding to amino acid residues L10-K60. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction