Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UTP14A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575335
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575335 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575335 Supplier Invitrogen™ Supplier No. PA575335
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

UTP14A, a related protein, may also be required for ribosome biogenesis, but not necessarily in a male-specific manner. The gene encoding UTP14C maps to human chromosome 13, which houses over 400 genes, such as BRCA2 and RB1, and comprises nearly 4% of the human genome. As with most chromosomes, polysomy of part or all of chromosome 13 is deleterious to development and decreases the odds of survival. Trisomy 13, also known as Patau syndrome, is deadly and the few who survive past one year suffer from permanent neurologic defects, difficulty eating and vulnerability to serious respiratory infections.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UTP14A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide; pH 7.2
Gene UTP14A
Gene Accession No. Q640M1, Q9BVJ6
Gene Alias 2700066J21Rik; Antigen NY-CO-16; dJ537K23.3; Jsd-like X-linked protein; JsdX; Juvenile spermatogonial depletion-like X-linked protein; Kiaa0266; mKIAA0266; NYCO16; NY-CO-16; SDCCAG16; Serologically defined colon cancer antigen 16; T25628; U3 small nucleolar RNA-associated protein 14 homolog A; UTP14, U3 small nucleolar ribonucleoprotein, homolog A; UTP14, U3 small nucleolar ribonucleoprotein, homolog A (yeast); UTP14A; UTP14A small subunit (SSU) processome component; UTP14A small subunit processome component
Gene Symbols UTP14A
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the N-terminual of Human UTP14A.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10813, 72554
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.