Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UTP14C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561468
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561468 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561468 Supplier Invitrogen™ Supplier No. PA561468
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ARMMERMSLK HQNSGKWAKS KAIMAKYDLE ARQAMQEQLA KNKELTQKLQ VASESEEE Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 97%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen UTP14C
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UTP14C
Gene Accession No. Q5TAP6
Gene Alias 2700066J21Rik; KIAA0266; U3 small nucleolar RNA-associated protein 14 homolog C; UTP14, small subunit processome component homolog C (S. cerevisiae); UTP14, U3 small nucleolar ribonucleoprotein, homolog C; UTP14B; UTP14C
Gene Symbols UTP14C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UTP14C. Recombinant protein control fragment (Product #RP-99596).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9724
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.