Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UTP15 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | UTP15 |
---|---|
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
UTP15 Polyclonal specifically detects UTP15 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
UTP15 | |
Polyclonal | |
Rabbit | |
Human | |
84135 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LWDIPNSKEILTFKEHSDYVRCGCASKLNPDLFITGSYDHTVKMFDARTSESVLSVEHGQPVESVLLFPSGGLLVSAG | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
U3 small nucleolar ribonucleoprotein, homolog (yeast), U3 small nucleolar RNA-associated protein 15 homolog, UTP15, U3 small nucleolar ribonucleoprotein, homolog (S. cerevisiae) | |
UTP15 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title