Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UTP15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113210
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113210 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113210 Supplier Invitrogen™ Supplier No. PA5113210
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UTP15 Polyclonal Antibody for Western Blot, ELISA

Ribosome biogenesis factor. Involved in nucleolar processing of pre-18S ribosomal RNA. Required for optimal pre-ribosomal RNA transcription by RNA polymerase I. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen UTP15
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene UTP15
Gene Accession No. Q8TED0
Gene Alias AW544865; LOW QUALITY PROTEIN: U3 small nucleolar RNA-associated protein 15 homolog; NET21; RGD1310992; Src-associated protein SAW; U3 small nucleolar RNA-associated protein 15 homolog; U3 small nucleolar RNA-associated protein 15 homolog; LOW QUALITY PROTEIN: U3 small nucleolar RNA-associated protein 15 homolog; UniProtKB:Q8TED0}; UTP15; utp15 {ECO:0000250; UTP15 small subunit processome component; UTP15 SSU processome component; UTP15, small subunit processome component; UTP15, U3 small nucleolar ribonucleoprotein, homolog; UTP15, U3 small nucleolar ribonucleoprotein, homolog (S. cerevisiae); UTP15, U3 small nucleolar ribonucleoprotein, homolog (yeast)
Gene Symbols UTP15
Host Species Rabbit
Immunogen Recombinant Human U3 small nucleolar RNA-associated protein 15 homolog protein (234-518AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84135
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.