Learn More
UTP15 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305119A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Orangutan
Involved in nucleolar processing of pre-18S ribosomal RNA.Specifications
UTP15 | |
Polyclonal | |
Unconjugated | |
UTP15 | |
NET21; U3 small nucleolar RNA-associated protein 15 homolog; Src-associated protein SAW; UTP15 SSU processome component; UTP15, U3 small nucleolar ribonucleoprotein, homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84135 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q8TED0 | |
UTP15 | |
Between 1 and 50 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.