Learn More
UTP15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | UTP15 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-1564
![]() ![]() |
Proteintech Group Inc
174161AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Product No. 17416-1-AP is a rabbit polyclonal antibody raised against the full-length of human UTP15. It specifically recognizes 58 and 37 kDa isoforms.
Involved in nucleolar processing of pre-18S ribosomal RNA.Specifications
UTP15 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
A2RRU3, Q8C7V3, Q8TED0 | |
105372, 310019, 84135 | |
UTP15 Fusion Protein Ag11161 | |
Primary | |
-20°C | |
UTP15 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
NET21, UTP15 | |
Utp15 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.