Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UTP18 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297492F
Description
UTP18 Polyclonal specifically detects UTP18 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
UTP18 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
CGI-48, U3 small nucleolar RNA-associated protein 18 homolog, UTP18, small subunit (SSU) processome component, homolog (yeast), WD repeat domain 50, WD repeat-containing protein 50, WDR50 | |
Produced in rabbits immunized with E. coli-derived Human UTP18 fragment. | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
51096 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
FITC | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction