Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UTP18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541860
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541860 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541860 Supplier Invitrogen™ Supplier No. PA541860
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PARPSAAAAA IAVAAAEEER RLRQRNRLRL EEDKPAVERC LEELVFGDVE Sequence homology: Dog: 93%; Human: 100%; Pig: 86%; Rabbit: 92%.

UTP18 is involved in nucleolar processing of pre-18S ribosomal RNA and ribosome assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UTP18
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene UTP18
Gene Accession No. Q9Y5J1
Gene Alias 6230425C22Rik; 6430513M13; CDABP0061; CGI-48; U3 small nucleolar RNA-associated protein 18 homolog; Utp18; UTP18 small subunit (SSU) processome component homolog; UTP18 small subunit processome component; UTP18, small subunit (SSU) processome component, homolog; UTP18, small subunit (SSU) processome component, homolog (yeast); UTP18, small subunit processome component; WD repeat domain 50; WD repeat-containing protein 50; Wdr50
Gene Symbols UTP18
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human UTP18 (aa 51-100).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51096
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.