Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UTP18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562596
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562596 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562596 Supplier Invitrogen™ Supplier No. PA562596
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HEDSGDSEVE NEAKGNFPPQ KKPVWVDEED EDEEMVDMMN NRFRKDMMKN ASESKLSKDN LKKRLKEEFQ HAMGGVPAWA ETTKRKT Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 81%.

UTP18 is involved in nucleolar processing of pre-18S ribosomal RNA and ribosome assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UTP18
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UTP18
Gene Accession No. Q9Y5J1
Gene Alias 6230425C22Rik; 6430513M13; CDABP0061; CGI-48; U3 small nucleolar RNA-associated protein 18 homolog; Utp18; UTP18 small subunit (SSU) processome component homolog; UTP18 small subunit processome component; UTP18, small subunit (SSU) processome component, homolog; UTP18, small subunit (SSU) processome component, homolog (yeast); UTP18, small subunit processome component; WD repeat domain 50; WD repeat-containing protein 50; Wdr50
Gene Symbols UTP18
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UTP18. Recombinant protein control fragment (Product #RP-93228).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51096
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.