Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UTP18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567632
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567632 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567632 Supplier Invitrogen™ Supplier No. PA567632
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UTP18 Polyclonal Antibody for Western Blot

UTP18 is involved in nucleolar processing of pre-18S ribosomal RNA and ribosome assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UTP18
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene UTP18
Gene Accession No. Q9Y5J1
Gene Alias 6230425C22Rik; 6430513M13; CDABP0061; CGI-48; U3 small nucleolar RNA-associated protein 18 homolog; Utp18; UTP18 small subunit (SSU) processome component homolog; UTP18 small subunit processome component; UTP18, small subunit (SSU) processome component, homolog; UTP18, small subunit (SSU) processome component, homolog (yeast); UTP18, small subunit processome component; WD repeat domain 50; WD repeat-containing protein 50; Wdr50
Gene Symbols UTP18
Host Species Rabbit
Immunogen Synthesized peptide derived from human UTP18.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51096
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.