Learn More
Invitrogen™ UTP18 Polyclonal Antibody

Description
Immunogen sequence: RRLRQRNRLR LEEDKPAVER CLEELVFGDV ENDEDALLRR LRGPRVQEHE DSGDSEVENE AKGNFPPQKK PVWVDEEDED EEMVDMMNNR FRKDMMKNAS ESKLSKDNLK KRLKEEFQHA MGGVPAWAET TKRKTSSDDE SEEDEDDLLQ RTGNFISTST SLPRGILKMK NCQHANAERP TVARISSVQF H; Positive Samples: HeLa, Jurkat, Mouse brain, Mouse kidney, Mouse liver; Cellular Location: Nucleus, nucleolus.
Specifications
Specifications
| Antigen | UTP18 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.77 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | UTP18 |
| Gene Accession No. | Q5SSI6, Q9Y5J1 |
| Gene Alias | 6230425C22Rik; 6430513M13; CDABP0061; CGI-48; U3 small nucleolar RNA-associated protein 18 homolog; Utp18; UTP18 small subunit (SSU) processome component homolog; UTP18 small subunit processome component; UTP18, small subunit (SSU) processome component, homolog; UTP18, small subunit (SSU) processome component, homolog (yeast); UTP18, small subunit processome component; WD repeat domain 50; WD repeat-containing protein 50; Wdr50 |
| Gene Symbols | UTP18 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.