Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UTP18/WDR50 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501560677
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-0677 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-0677 Supplier Bethyl Laboratories, Inc Supplier No. A301551AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

UTP18 is involved in nucleolar processing of pre-18S ribosomal RNA.

Specifications

Antigen UTP18/WDR50
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene UTP18
Gene Accession No. Q9Y5J1
Gene Alias CGI-48, UTP18 small subunit (SSU) processome component homolog, UTP18 small subunit (SSU) processome component homolog (yeast), UTP18 small subunit processome component, UTP18, small subunit (SSU) processome component, homolog, WD repeat domain 50, WD repeat-containing protein 50, WDR50
Gene Symbols UTP18
Host Species Rabbit
Immunogen Between 75 and 125
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51096
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.