Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UTP20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565554
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565554 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565554 Supplier Invitrogen™ Supplier No. PA565554
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FETLSDFESGL KYITDVVKLN AFDQRHLDDI NFDVRFETFQ TITSYIKEMQ IVDVNYLIPV MHNCFYNLEL GDMSLSDNAS MC Highest antigen sequence identity to the following orthologs - mouse 81%, rat 81%.

Isoform 1 may function as a nuclear receptor coactivator, enhancing transcription through other coactivators such as NCOA6 and CITED1. Isoform 2, functions as a transcriptional repressor, modulating transcriptional activities of coactivators including isoform 1, NCOA6 and CITED1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UTP20
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UTP20
Gene Accession No. O75691
Gene Alias 1A6/DRIM; 3830408P06Rik; AA617408; down regulated in metastasis; down-regulated in metastasis protein; DRIM; mDRIM; mDRIN; NNP73; novel nucleolar protein 73; Protein Key-1A6; RGD1560606; Small subunit processome component 20 homolog; UTP20; UTP20 small subunit (SSU) processome component; UTP20 small subunit processome component; UTP20, small subunit (SSU) processome component, homolog; UTP20, small subunit (SSU) processome component, homolog (yeast); UTP20, small subunit processome component
Gene Symbols UTP20
Host Species Rabbit
Immunogen Recombinant Human UTP20. Recombinant protein control fragment (Product #RP-106228).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27340
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.