Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UTP23 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | UTP23 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
UTP23 Polyclonal specifically detects UTP23 in Human samples. It is validated for Western Blot.Specifications
| UTP23 | |
| Polyclonal | |
| Rabbit | |
| NP_115710 | |
| 84294 | |
| Synthetic peptide directed towards the C terminal of human UTP23The immunogen for this antibody is UTP23. Peptide sequence SGPNPLSCLKKKKKAPDTQSSASEKKRKRKRIRNRSNPKVLSEKQNAEGE. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| C8orf53, chromosome 8 open reading frame 53, MGC14595, rRNA-processing protein UTP23 homolog, UTP23, small subunit (SSU) processome component, homolog (yeast) | |
| UTP23 | |
| IgG | |
| 28 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title