Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UTP23 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113401
Description
UTP23 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Specifications
| UTP23 | |
| Polyclonal | |
| Unconjugated | |
| UTP23 | |
| 1700010I21Rik; AI662478; BB238373; C8orf53; D530033C11Rik; RGD1307179; rRNA-processing protein UTP23 homolog; UTP23; UTP23, small subunit (SSU) processome component, homolog; UTP23, small subunit (SSU) processome component, homolog (yeast); UTP23, small subunit processome component | |
| Rabbit | |
| Protein G | |
| RUO | |
| 78581, 84294 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 3.5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9BRU9, Q9CX11 | |
| UTP23 | |
| Recombinant Human rRNA-processing protein UTP23 homolog protein (1-249AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction