Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UTP3 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP119108B
Description
UTP3 Polyclonal antibody specifically detects UTP3 in Human samples. It is validated for Western BlotSpecifications
UTP3 | |
Polyclonal | |
Western Blot | |
Charged amino acid-rich leucine zipper 1, CRL1charged amino acid rich leucine zipper 1 homolog, CRLZ1UTP3 homolog, disrupter of silencing 10, Disrupter of silencing SAS10, FLJ23256, SAS10DKFZp761F222, something about silencing protein 10, UTP3, small subunit (SSU) processome component, homolog (S. cerevisiae) | |
The immunogen recognized by this antibody maps to a region between residue 429 and 479 of human something about silencing protein 10 using the numbering given in entry NP_065101.1 (GeneID 57050). | |
0.1 mL | |
Chromatin Modifiers | |
57050 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Biotin | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction