Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UTP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565299
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565299 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565299 Supplier Invitrogen™ Supplier No. PA565299
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LNYCSNISFYL ILKARRVPAH GHPVIERLVT YRNLINKLSV VDQKLSSEIR HLLTLKDDAV KKELIPKAKS TKPK Highest antigen sequence identity to the following orthologs - mouse 91%, rat 88%.

Essential for gene silencing: has a role in the structure of silenced chromatin. Plays a role in the developing brain. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen UTP3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Utp3
Gene Accession No. Q9NQZ2
Gene Alias 2400011K09Rik; AW557551; C87704; charged amino acid rich leucine zipper 1; charged amino acid rich leucine zipper 1 homolog; charged amino acid-rich leucine zipper 1; Crl1; Crl-1; Crlz1; disrupter of silencing 10; disrupter of silencing SAS10; Sas10; something about silencing protein 10; UTP3; UTP3 homolog; UTP3, small subunit (SSU) processome component, homolog (S. cerevisiae); UTP3, small subunit processome component; UTP3, small subunit processome component homolog (S. cerevisiae)
Gene Symbols Utp3
Host Species Rabbit
Immunogen Recombinant Human UTP3. Recombinant protein control fragment (Product #RP-104735).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57050
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.