Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UTP6 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP130467AF750
Description
UTP6 Polyclonal antibody specifically detects UTP6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
UTP6 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
C17orf40, chromosome 17 open reading frame 40, HCA66hepatocellular carcinoma associated antigen 66, Hepatocellular carcinoma-associated antigen 66, MHAT, Multiple hat domains protein, U3 small nucleolar RNA-associated protein 6 homolog, UTP6, small subunit (SSU) processome component, homolog (yeast) | |
The immunogen for this product maps to a region between residue 565 and 597 of human hepatocellular carcinoma associated antigen 66 using the numbering given in entry NP_060898.2 (GeneID 55813). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
55813 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction