Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UTP6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555272
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555272 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555272 Supplier Invitrogen™ Supplier No. PA555272
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ARQLFLRALR FHPECPKLYK EYFRMELMHA EKLRKEKEEF EKASMDVENP DYSEEILKGE LAWIIYKNSV SIIKGAEFHV SLLSIAQLFD FAKDLQKEIY DDLQALHTDD PLTWDYVARR ELEIESQTEE QPTTKQAKAV Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 86%.

Involved in nucleolar processing of pre-18S ribosomal RNA. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen UTP6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UTP6
Gene Accession No. Q8VCY6, Q9NYH9
Gene Alias 4732497O03Rik; C17orf40; HCA66; hepatocellular carcinoma associated antigen 66; hepatocellular carcinoma-associated antigen 66; Mhat; Multiple hat domains protein; RGD1311017; U3 small nucleolar RNA-associated protein 6 homolog; Utp6; UTP6 small subunit processome component; UTP6, small subunit (SSU) processome component, homolog; UTP6, small subunit (SSU) processome component, homolog (yeast); UTP6, small subunit processome component
Gene Symbols UTP6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UTP6. Recombinant protein control fragment (Product #RP-93203).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216987, 360574, 55813
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.