Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UTP6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116771
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116771 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116771 Supplier Invitrogen™ Supplier No. PA5116771
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HT-29, HeLa, Mouse lung, Mouse thymus, Mouse spleen, Rat lung, Rat thymus Immunogen sequence: MAEIIQERIE DRLPELEQLE RIGLFSHAEI KAIIKKASDL EYKIQRRTLF KEDFINYVQY EINLLELIQR RRTRIGYSFK KDEIENSIVH RVQGVFQRAS AKWKDDVQLW LSYVAFCKKW ATKTRLSKVF SAMLAIHSNK PALWIMAAKW EMEDRLSSES ARQLFLRALR FHPECPKLYK EYFRMELMHA EKLRKEKEEF EKASMDVENP DYSEEILKGE.

Involved in nucleolar processing of pre-18S ribosomal RNA. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen UTP6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene UTP6
Gene Accession No. Q8VCY6, Q9NYH9
Gene Alias 4732497O03Rik; C17orf40; HCA66; hepatocellular carcinoma associated antigen 66; hepatocellular carcinoma-associated antigen 66; Mhat; Multiple hat domains protein; RGD1311017; U3 small nucleolar RNA-associated protein 6 homolog; Utp6; UTP6 small subunit processome component; UTP6, small subunit (SSU) processome component, homolog; UTP6, small subunit (SSU) processome component, homolog (yeast); UTP6, small subunit processome component
Gene Symbols UTP6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human UTP6 (NP_0608982).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216987, 360574, 55813
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.