Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UTRN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553873
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553873 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553873 Supplier Invitrogen™ Supplier No. PA553873
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GELDKQLTTY LTDRIDAFQV PQEAQKIQAE ISAHELTLEE LRRNMRSQPL TSPESRTARG GSQMDVLQRK LREVSTKFQL FQKPANFEQR MLDCKRVLDG VKAELHVLDV KDVDPDVIQT HLDKCMKLYK TLSEVKLEVE TVI Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.

The UTRN gene encodes the utrophin protein, which is closely related to dystrophin in structure and function, playing a crucial role in maintaining the integrity of muscle fiber membranes. Located on chromosome 6, utrophin is expressed widely in fetal tissues but is replaced by dystrophin in mature muscle fibers; however, it remains present in adult tissues such as the neuromuscular junction and blood vessels. Utrophin's primary role is to stabilize the cytoskeleton and membrane linkages, forming part of the protein complex responsible for structural support in muscle cells. This protein has garnered significant attention in the context of muscular dystrophies, particularly Duchenne Muscular Dystrophy (DMD), where utrophin upregulation is considered a potential therapeutic strategy to compensate for the lack of functional dystrophin. It helps to ameliorate muscle pathology by mimicking the stabilizing effects of dystrophin, thus preserving muscle function and reducing damage. Research on enhancing utrophin expression is ongoing, aiming to provide an effective treatment modality for DMD and other dystrophin-associated disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UTRN
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UTRN
Gene Accession No. P46939
Gene Alias DMDL; DRP; DRP1; DRP-1; Dystrophin-related protein 1; UTRN; Utrophin
Gene Symbols UTRN
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UTRN. Recombinant protein control fragment (Product #RP-101029).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7402
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.