Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Utrophin A Rabbit anti-Human, Mouse, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABN173925UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN1739-25UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1739-25UG Supplier MilliporeSigma Supplier No. ABN173925UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Utrophin A Antibody, Cat. No. ABN1739, is a highly specific rabbit polyclonal antibody that targets Utrophin and has been tested in Immunofluorescence, Immunohistochemistry (Paraffin), and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Utrophin A
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in PBS with 0.02% sodium azide.
Gene Alias UTRN A
Gene Symbols UTRN
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the first 26 amino acids from the N-terminal region of murine utrophin.
Molecular Weight of Antigen 394.46 kDa calculated.
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects utrophin in human and mouse muscle tissue. It targets an epitope with in the first 26 amino acids from the N-terminal region.
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.