Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Invitrogen™ UTS2 Polyclonal Antibody
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110561
Description
Immunogen sequence: APHEDARLTP EELERASLLQ ILPEMLGAER GDILRKADSS TNIFNPRGNL RKFQDFSGQD PNILLSHLLA RIWKPYKKRE TPDCFWKYCV.
Urotensin II is an active cyclic heptapeptide absolutely conserved from lamprey to human. The active peptide acts as a vasoconstrictor and is expressed only in brain tissue. Despite the gene family name similarity, this gene is not homologous to urocortin, a member of the sauvagine/corticotropin releasing factor/urotensin I family. Most of the proprotein is cleaved to make the mature peptide.
            Specifications
| UTS2 | |
| Polyclonal | |
| Unconjugated | |
| UTS2 | |
| prepro U-II; prepro-UII; PRO1068; Ucn2; UII; U-II; UNQ525/PRO1068; urocortin 2; urotensin 2; urotensin II; Urotensin-2; urotensin-II; UTS2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10911, 24111 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | 
| ELISA, Western Blot | |
| 2.53 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O95399, Q9QZQ3 | |
| UTS2 | |
| A synthetic peptide corresponding to a sequence within amino acids 35-124 of human UTS2 (NP_0067771). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG | 
Safety and Handling
                                                    
                                                    WARNING: Cancer - www.P65Warnings.ca.gov 
                                                
                                            
                                        
                                    
                                
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction