Learn More
UTS2B Polyclonal Antibody

Description
UTS2B Polyclonal Antibody for IHC
Specifications
Specifications
| Antigen | UTS2B |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | UTS2B |
| Gene Accession No. | Q765I0 |
| Gene Alias | Gm538; prepro-URP; U2B; UIIB; U-IIB; urotensin 2 domain containing; urotensin 2 domain containing protein; urotensin 2 domain-containing protein; urotensin 2B; Urotensin IIB; urotensin II-related peptide; urotensin-2 domain-containing protein; Urotensin-2B; urotensin-IIB; Urp; Uts2b; Uts2d |
| Gene Symbols | UTS2B |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.