Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UTS2B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987792
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7792 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7792 Supplier Bioss Supplier No. BS7511R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UTS2B (Urotensin 2B) is a Protein Coding gene. Among its related pathways are Signaling by GPCR and RET signaling. Gene Ontology (GO) annotations related to this gene include hormone activity and G protein-coupled receptor binding. Also binds urotensin II and produces hypotension.

Specifications

Antigen UTS2B
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene UTS2B
Gene Accession No. Q765I0
Gene Alias prepro-URP; U2B; UIIB; U-IIB; urotensin 2 domain containing; urotensin 2B; Urotensin IIB; Urotensin II-related peptide; Urotensin-2 domain-containing protein; Urotensin-2B; URP; UTS2B; UTS2D
Gene Symbols UTS2B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human UTS2D.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 257313
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.